Assay Details
| Gene Symbol: Chmp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025519.2 | NP_079795.1 | 2 - 3 | 432 | 82 | |
| GenBank mRNA | AK009273.1 | - | 2 - 3 | 432 | 82 | |
| AK010707.1 | - | 2 - 3 | 499 | 82 | ||
| AK160189.1 | - | 2 - 3 | 426 | 82 | ||
| BC115983.1 | - | 2 - 3 | 422 | 40105850 | 82 | |
| BC117538.1 | - | 2 - 3 | 422 | 40105849 | 82 | |
More Information
Set Membership: |
|

Chmp4c
Genomic Map