Assay Details
| Gene Symbol: Nudt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025529.3 | NP_079805.1 | 2 - 3 | 397 | 97 | |
| GenBank mRNA | AK002605.1 | - | 2 - 3 | 335 | 97 | |
| AK009700.1 | - | 2 - 3 | 335 | 97 | ||
| AK045197.1 | - | 2 - 3 | 398 | 97 | ||
| AK170076.1 | - | 2 - 3 | 337 | 97 | ||
| BC056443.1 | - | 2 - 3 | 337 | 6822891 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
thiamine-pyrophosphatase activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity
metal ion binding
biological_process
thiamine-pyrophosphatase activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31920557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31920558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31920556 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | NM_025529.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | AK002605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | AK009700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | AK045197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | AK170076.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | NM_025529.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | AK002605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | AK009700.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | AK045197.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490388 | AK170076.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 167453 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102869 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | NM_025529.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | AK002605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | AK009700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | AK045197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | AK170076.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | NM_025529.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | AK002605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | AK009700.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | AK045197.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490388 | AK170076.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | NM_025529.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | AK002605.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | AK009700.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | AK045197.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | AK170076.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | NM_025529.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | AK002605.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | AK009700.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | AK045197.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490388 | AK170076.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31920561 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102869 | - | - |
| Whole Mouse Genome G4122A | A_51_P490388 | NM_025529.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | AK002605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | AK009700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | AK045197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | AK170076.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | NM_025529.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | AK002605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | AK009700.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | AK045197.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P490388 | AK170076.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136990 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102869 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14151853 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14151825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14151834 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14151807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14151852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14151810 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14151848 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14151854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31920557 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31920556 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31920558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31920622 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14151815 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31920552 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10460287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10460286 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450111_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10460285 | - | - |
| Mouse Genome 430 2.0 Array | 1450111_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450111_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66387_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66387_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900000272.mm.1 | NM_025529.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000272.mm.1 | NM_025529.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000272.mm.1 | TC1900000024.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900000271.mm.1 | NM_025529.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000271.mm.1 | NM_025529.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900000271.mm.1 | TC1900000024.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450111_PM_a_at | - | - |
| Mouse Expression Set 430 | 1450111_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450111_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450111_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450111_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450111_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 356794 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3710441 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003710441 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2617305 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2617305 | - | - |
| mouse-6 v1.1 (Illumina) | scl52798.4.1_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3710441 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52798.4.1_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3710441 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2617305 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2617305 | - | - |

Nudt8
Genomic Map