Assay Details
| Gene Symbol: Mad2l1bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025649.3 | NP_079925.2 | 2 - 3 | 373 | 66 | |
| GenBank mRNA | AK002377.1 | - | 2 - 3 | 359 | 66 | |
| AK004224.1 | - | 2 - 3 | 327 | 66 | ||
| AK007974.1 | - | 2 - 3 | 325 | 66 | ||
| AK156569.1 | - | 2 - 3 | 66 | |||
| AK220322.1 | - | 2 - 3 | 66 | |||
| BC005768.1 | - | 2 - 3 | 2649017 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31205406 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31205408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31205409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31205407 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P425632 | AK002377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P425632 | AK002377.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827873 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150972 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166667 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132434 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164596 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P425632 | AK002377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P425632 | AK002377.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P425632 | AK002377.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P425632 | AK002377.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205386 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205356 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31205403 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132434 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03324301 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166667 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164596 | - | - |
| Whole Mouse Genome G4122A | A_51_P425632 | AK002377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P425632 | AK002377.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122475 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166667 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132434 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164596 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13319771 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13319766 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13213109 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13319774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13421359 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13319772 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13213111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13319768 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13213117 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31205406 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31205408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31205409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31205407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31205325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13103213 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13430062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13581270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13405221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13506905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13103216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13213109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13178717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31205363 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13421359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13103212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13468874 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10451222 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451221 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415720_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451220 | - | - |
| Mouse Genome 430 2.0 Array | 1415720_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1415720_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66591_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66591_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700020328.mm.1 | NM_025649.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020328.mm.1 | NM_025649.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020328.mm.1 | TC1700002165.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 95565_at | - | - |
| Murine Genome U74v2 Set | 95565_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415720_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415720_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415720_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415720_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 879546 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6020364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6020364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6130280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221616 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2669087 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1221616 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2669087 | - | - |
| mouse-6 v1.1 (Illumina) | scl0076588.1_15-S | - | - |
| mouse-6 v1.1 (Illumina) | scl49875.3_8-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130280 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0076588.1_15-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49875.3_8-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130280 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2815222 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1221616 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2669087 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221616 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2669087 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2815222 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl49875.3_8-S | - | - |

Mad2l1bp
Genomic Map