Assay Details
| Gene Symbol: Tmigd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025655.2 | NP_079931.1 | 2 - 3 | 374 | 83 | |
| GenBank mRNA | AK008028.1 | - | 3 - 4 | 435 | 83 | |
| AK009118.1 | - | 2 - 3 | 374 | 83 | ||
| BC022683.1 | - | 2 - 3 | 341 | 83 | ||
| BC061082.1 | - | 2 - 3 | 364 | 30311543 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmigd1
Genomic Map