Assay Details
| Gene Symbol: Ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025660.2 | NP_079936.1 | 7 - 8 | 1287 | 73 | |
| GenBank mRNA | AK011592.1 | - | 7 - 8 | 1287 | 73 | |
| BC050755.1 | - | 7 - 8 | 1275 | 6772058 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32747582 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32747584 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32747583 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217706 | BC050755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217706 | NM_025660.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217706 | AK011592.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217706 | BC050755.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217706 | NM_025660.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P217706 | AK011592.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830959 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357032 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485303 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927840 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141766 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 540101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537725 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170286 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110292 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139252 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217706 | BC050755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217706 | NM_025660.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217706 | AK011592.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217706 | BC050755.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217706 | NM_025660.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P217706 | AK011592.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747582 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747579 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747561 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32747571 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139252 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09460467 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09460460 | - | - |
| Whole Mouse Genome G4122A | A_51_P217706 | BC050755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217706 | NM_025660.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217706 | AK011592.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217706 | BC050755.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217706 | NM_025660.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P217706 | AK011592.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129127 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139252 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15056628 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15056640 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15056631 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32747584 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32747582 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32747583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32747507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747524 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14913408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14895567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15041027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15198452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747555 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14969193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14895579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14737250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14737253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14744545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32747531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15041016 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10607338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607339 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607344 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10607337 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449914_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10607336 | - | - |
| Mouse Genome 430 2.0 Array | 1449914_at | - | - |
| Mouse Genome 430 2.0 Array | 1449914_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66611_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66611_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00020012.mm.1 | NM_025660.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020012.mm.1 | NM_025660.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00020012.mm.1 | TC0X00003185.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449914_at | - | - |
| Mouse Expression Set 430 | 1449914_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449914_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449914_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449914_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449914_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 404916 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4150601 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4150601 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004150601 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2688533 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2688533 | - | - |
| mouse-6 v1.1 (Illumina) | scl53720.7.1_159-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4150601 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53720.7.1_159-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688533 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688533 | - | - |

Ribc1
Genomic Map