Assay Details
| Gene Symbol: Dcbld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025705.3 | NP_079981.2 | 7 - 8 | 867 | 92 | |
| GenBank mRNA | AK014521.1 | - | 7 - 8 | 679 | 92 | |
| AK016485.1 | - | 7 - 8 | 867 | 92 | ||
| BC026771.1 | - | 6 - 7 | 655 | 4014778 | 92 | |
More Information
Set Membership: |
|

Dcbld1
Genomic Map