Assay Details
| Gene Symbol: Pcyox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025823.4 | NP_080099.1 | 3 - 4 | 554 | 96 | |
| GenBank mRNA | AK004799.2 | - | 3 - 4 | 509 | 96 | |
| AK004840.1 | - | 3 - 4 | 509 | 96 | ||
| AK031585.1 | - | 3 - 4 | 510 | 96 | ||
| AK033373.1 | - | 3 - 4 | 510 | 96 | ||
| AK049412.1 | - | 3 - 4 | 507 | 96 | ||
| AK134528.1 | - | 3 - 4 | 555 | 96 | ||
| AK145366.1 | - | 3 - 4 | 514 | 96 | ||
| AK147190.1 | - | 3 - 4 | 522 | 96 | ||
| AK159981.1 | - | 3 - 4 | 510 | 96 | ||
| AK161616.1 | - | 3 - 4 | 510 | 96 | ||
| AK169209.1 | - | 3 - 4 | 510 | 96 | ||
| AK173058.1 | - | 3 - 4 | 476 | 96 | ||
| BC006056.1 | - | 1 - 2 | 165 | 96 | ||
| BC028308.1 | - | 3 - 4 | 524 | 3486530 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcyox1
Genomic Map