Assay Details
| Gene Symbol: Lonp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025827.3 | NP_080103.1 | 6 - 7 | 1123 | 84 | |
| GenBank mRNA | AK004843.1 | - | 6 - 7 | 1037 | 84 | |
| AK050231.1 | - | 6 - 7 | 1032 | 84 | ||
| AK151103.1 | - | 6 - 7 | 1069 | 84 | ||
| AK168791.1 | - | 6 - 7 | 1069 | 84 | ||
| AK168956.1 | - | 6 - 7 | 1069 | 84 | ||
| AK169656.1 | - | 1037 | 84 | |||
| BC049090.1 | - | 6 - 7 | 1059 | 6401542 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lonp2
Genomic Map