Assay Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025850.2 | NP_080126.1 | 5 - 6 | 536 | 65 | |
| GenBank mRNA | AK005694.1 | - | 5 - 6 | 546 | 65 | |
| AY819025.1 | - | 5 - 6 | 566 | 65 | ||
| BC046278.1 | - | 5 - 6 | 536 | 6310911 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fank1
Genomic Map