Assay Details
| Gene Symbol: Fank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025850.2 | NP_080126.1 | 6 - 7 | 606 | 77 | |
| GenBank mRNA | AK005694.1 | - | 6 - 7 | 616 | 77 | |
| AK017071.1 | - | 6 - 7 | 630 | 77 | ||
| AY819025.1 | - | 6 - 7 | 636 | 77 | ||
| BC046278.1 | - | 6 - 7 | 606 | 6310911 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fank1
Genomic Map