Assay Details
| Gene Symbol: Rassf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025886.3 | NP_080162.3 | 1 - 2 | 76 | 66 | |
| GenBank mRNA | AK010305.1 | - | 1 - 2 | 103 | 66 | |
| CK617449.1 | - | 1 - 2 | 76 | 66 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_025886.3 transcripts.
Set Membership: |
|

Rassf7
Genomic Map