Assay Details
| Gene Symbol: Rassf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025886.3 | NP_080162.3 | 2 - 3 | 215 | 68 | |
| GenBank mRNA | AK002243.1 | - | 2 - 3 | 155 | 68 | |
| AK010305.1 | - | 2 - 3 | 242 | 68 | ||
| BC011131.1 | - | 1 - 2 | 142 | 4206694 | 68 | |
| CK617449.1 | - | 2 - 3 | 215 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rassf7
Genomic Map