Assay Details
| Gene Symbol: Rassf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025886.3 | NP_080162.3 | 5 - 6 | 1137 | 54 | |
| GenBank mRNA | AK010305.1 | - | 5 - 6 | 1163 | 54 | |
| AW990538.1 | - | 1 - 2 | 54 | |||
| BC011131.1 | - | 4 - 5 | 1064 | 4206694 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587470 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587469 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587471 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587468 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587561 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587560 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | AK002243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | AK010305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | AW990538.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | BC011131.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | NM_025886.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | AK002243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | AK010305.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | AW990538.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | BC011131.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P521176 | NM_025886.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226407 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134066 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | AK002243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | AK010305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | AW990538.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | BC011131.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | NM_025886.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | AK002243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | AK010305.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | AW990538.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | BC011131.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P521176 | NM_025886.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587449 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25587437 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134066 | - | - |
| Whole Mouse Genome G4122A | A_51_P521176 | AK002243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | AK010305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | AW990538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | BC011131.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | NM_025886.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | AK002243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | AK010305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | AW990538.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | BC011131.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P521176 | NM_025886.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105454 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134066 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06123599 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06123570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06123602 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06123583 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06123620 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587471 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587468 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587561 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587470 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25587469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25587468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06123614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06123571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06123594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06123620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06123569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25587495 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558843 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418924_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558840 | - | - |
| Mouse Genome 430 2.0 Array | 1418924_at | - | - |
| Mouse Genome 430 2.0 Array | 1418924_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66985_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66985_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700018069.mm.1 | NM_025886.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018069.mm.1 | NM_025886.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018069.mm.1 | TC0700002043.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700018068.mm.1 | NM_025886.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018068.mm.1 | NM_025886.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700018068.mm.1 | TC0700002043.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418924_at | - | - |
| Mouse Expression Set 430 | 1418924_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418924_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418924_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418924_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418924_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 460372 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4920193 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1780692 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920193 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001780692 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2682778 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2778094 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2682778 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2778094 | - | - |
| mouse-6 v1.1 (Illumina) | scl31892.6.1_61-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00049.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920193 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1780692 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31892.6.1_61-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00049.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780692 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920193 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2778094 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2682778 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2682778 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2778094 | - | - |

Rassf7
Genomic Map