Assay Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025989.3 | NP_080265.2 | 2 - 3 | 222 | 86 | |
| GenBank mRNA | AK009323.1 | - | 2 - 3 | 223 | 86 | |
| AK009661.1 | - | 2 - 3 | 223 | 86 | ||
| AK009843.1 | - | 2 - 3 | 176 | 86 | ||
| BC119573.2 | - | 2 - 3 | 217 | 40057995 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gp2
Genomic Map