Assay Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025989.3 | NP_080265.2 | 10 - 11 | 1680 | 88 | |
| GenBank mRNA | AK009323.1 | - | 10 - 11 | 1681 | 88 | |
| AK009466.1 | - | 5 - 6 | 577 | 88 | ||
| AK009558.1 | - | 4 - 5 | 390 | 88 | ||
| AK009661.1 | - | 10 - 11 | 1681 | 88 | ||
| AK009843.1 | - | 9 - 10 | 1549 | 88 | ||
| BC119573.2 | - | 10 - 11 | 1675 | 40057995 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965881 | AK009661.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965881 | AK009466.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965881 | AK009558.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965881 | NM_025989.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965882 | AK009661.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965882 | AK009843.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965882 | AK009323.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965882 | AK009466.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965882 | AK009558.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 665250 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820095 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537370 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480715 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 70264 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141085 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103930 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178727 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965881 | AK009661.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965881 | AK009466.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965881 | AK009558.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965881 | NM_025989.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965882 | AK009661.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965882 | AK009843.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965882 | AK009323.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965882 | AK009466.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965882 | AK009558.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268391 | AK009661.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268391 | AK009466.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268391 | AK009558.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P268391 | NM_025989.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463768 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25463757 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178727 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06342194 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141085 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103930 | - | - |
| Whole Mouse Genome G4122A | A_51_P268391 | AK009661.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268391 | AK009466.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268391 | AK009558.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P268391 | NM_025989.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105923 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141085 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178727 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06601812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06601823 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06601809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06601818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06601817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25463744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06601809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06636040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06161336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463669 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06160290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05917277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06160299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06090486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05908220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05908214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06090479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06090482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06161337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25463663 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10567379 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567368 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567374 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567375 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567376 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567378 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567372 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567371 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567373 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567369 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567367 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567377 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449452_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10567366 | - | - |
| Mouse Genome 430 2.0 Array | 1449452_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1449452_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67133_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67133_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700034776.mm.1 | NM_025989.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700034776.mm.1 | NM_025989.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700034776.mm.1 | TC0700004182.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700034775.mm.1 | NM_025989.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700034775.mm.1 | NM_025989.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700034775.mm.1 | TC0700004182.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449452_PM_a_at | - | - |
| Mouse Expression Set 430 | 1449452_a_at | - | - |
| Murine Genome U74v2 Set | 165494_i_at | - | - |
| Murine Genome U74v2 Set | 165494_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449452_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449452_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449452_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449452_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5860563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2735775 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2735775 | - | - |
| mouse-6 v1.1 (Illumina) | scl30747.11.1_143-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30747.11.1_143-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2904435 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2904435 | - | - |

Gp2
Genomic Map