Assay Details
| Gene Symbol: Tomm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291155.1 | NP_001278084.1 | 4 - 5 | 677 | 71 | |
| NM_025996.5 | NP_080272.1 | 4 - 5 | 1056 | 71 | ||
| GenBank mRNA | AK011786.1 | - | 4 - 5 | 1056 | 71 | |
| AK017699.1 | - | 4 - 5 | 622 | 71 | ||
| AK075873.1 | - | 4 - 5 | 674 | 71 | ||
| AK168329.1 | - | 4 - 5 | 611 | 71 | ||
| BC018278.1 | - | 4 - 5 | 607 | 4187356 | 71 | |
| CX235437.1 | - | 4 - 5 | 596 | 71 | ||
More Information
Set Membership: |
|

Tomm34
Genomic Map