Assay Details
| Gene Symbol: Tomm34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291155.1 | NP_001278084.1 | 5 - 6 | 828 | 68 | |
| NM_025996.5 | NP_080272.1 | 5 - 6 | 1207 | 68 | ||
| GenBank mRNA | AK011786.1 | - | 5 - 6 | 1207 | 68 | |
| AK017699.1 | - | 5 - 6 | 773 | 68 | ||
| AK075873.1 | - | 5 - 6 | 825 | 68 | ||
| AK168329.1 | - | 5 - 6 | 762 | 68 | ||
| BC018278.1 | - | 5 - 6 | 758 | 4187356 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tomm34
Genomic Map