Assay Details
| Gene Symbol: Rnf219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026047.4 | NP_080323.3 | 2 - 3 | 265 | 76 | |
| GenBank mRNA | AK013314.1 | - | 2 - 3 | 244 | 76 | |
| AK020974.1 | - | 2 - 3 | 233 | 76 | ||
| BC029231.1 | - | 2 - 3 | 231 | 3156840 | 76 | |
| BI986775.1 | - | 2 - 3 | 265 | 76 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC029231.1 transcripts.
Set Membership: |
|

Rnf219
Genomic Map