Assay Details
| Gene Symbol: Gtf3c6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026113.4 | NP_080389.2 | 2 - 3 | 164 | 91 | |
| GenBank mRNA | AK005261.1 | - | 2 - 3 | 163 | 91 | |
| AK007816.1 | - | 2 - 3 | 165 | 91 | ||
| AK010514.1 | - | 2 - 3 | 164 | 91 | ||
| AK168070.1 | - | 2 - 3 | 163 | 91 | ||
| BC027260.1 | - | 1 - 2 | 80 | 91 | ||
| BC048446.1 | - | 2 - 3 | 170 | 6590885 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c6
Genomic Map