Assay Details
| Gene Symbol: Calm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007590.3 | NP_031616.1 | 1 - 2 | 189 | 64 | |
| GenBank mRNA | AK151610.1 | - | 1 - 2 | 153 | 64 | |
| AK152754.1 | - | 1 - 2 | 138 | 64 | ||
| AK153179.1 | - | 1 - 2 | 126 | 64 | ||
| AK169630.1 | - | 1 - 2 | 104 | 64 | ||
| BC050926.1 | - | 1 - 2 | 111 | 6411878 | 64 | |
| BE853985.1 | - | 1 - 2 | 207 | 64 | ||
| M19380.1 | - | 1 - 2 | 109 | 64 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK151610.1,AK152754.1,AK153179.1,BC050926.1,M19380.1,NM_007590.3 transcripts.
Set Membership: |
|

Calm3
Genomic Map