Assay Details
| Gene Symbol: Capn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110504.1 | NP_001103974.1 | 7 - 8 | 932 | 75 | |
| NM_007600.3 | NP_031626.1 | 7 - 8 | 984 | 75 | ||
| GenBank mRNA | AF021847.1 | - | 7 - 8 | 863 | 75 | |
| AF084459.1 | - | 6 - 7 | 850 | 75 | ||
| AK088547.1 | - | 7 - 8 | 904 | 75 | ||
| AK149090.1 | - | 3 - 4 | 332 | 75 | ||
| AK149706.1 | - | 7 - 8 | 924 | 75 | ||
| AK150084.1 | - | 7 - 8 | 923 | 75 | ||
| AK160940.1 | - | 7 - 8 | 924 | 75 | ||
| AK165258.1 | - | 7 - 8 | 943 | 75 | ||
| AK168063.1 | - | 7 - 8 | 923 | 75 | ||
| AK171405.1 | - | 7 - 8 | 887 | 75 | ||
| BC026138.1 | - | 7 - 8 | 899 | 4218148 | 75 | |
| BC050276.1 | - | 7 - 8 | 900 | 4923347 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Capn1
Genomic Map