Assay Details
| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007658.3 | NP_031684.3 | 4 - 5 | 734 | 71 | |
| GenBank mRNA | AK077609.1 | - | 4 - 5 | 351 | 71 | |
| AK077991.1 | - | 4 - 5 | 737 | 71 | ||
| AK142334.1 | - | 3 - 4 | 1009 | 71 | ||
| BC046296.1 | - | 4 - 5 | 353 | 6486343 | 71 | |
| U27323.1 | - | 4 - 5 | 320 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25a
Genomic Map