Assay Details
| Gene Symbol: Cdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252627.1 | NP_001239556.1 | 9 - 10 | 1459 | 64 | |
| NM_001252628.1 | NP_001239557.1 | 10 - 11 | 1549 | 64 | ||
| NM_007663.3 | NP_031689.1 | 10 - 11 | 1549 | 64 | ||
| GenBank mRNA | AF016271.1 | - | 10 - 11 | 1417 | 64 | |
| AJ609635.1 | - | 10 - 11 | 1416 | 64 | ||
| AK052636.1 | - | 10 - 11 | 1352 | 64 | ||
| AK093905.1 | - | 10 - 11 | 1347 | 64 | ||
| AK164546.1 | - | 9 - 10 | 1460 | 64 | ||
| BC015251.1 | - | 10 - 11 | 1397 | 4235757 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh16
Genomic Map