Assay Details
| Gene Symbol: Cirbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007705.2 | NP_031731.1 | 1 - 2 | 74 | 82 | |
| GenBank mRNA | AK014655.1 | - | 1 - 2 | 77 | 82 | |
| AK132584.1 | - | 1 - 2 | 77 | 82 | ||
| BC075699.1 | - | 1 - 2 | 64 | 30656047 | 82 | |
| D78135.1 | - | 1 - 2 | 74 | 82 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK014655.1,AK132584.1,BC075699.1,D78135.1,NM_007705.2 transcripts.
Set Membership: |
|

Cirbp
Genomic Map