Assay Details
| Gene Symbol: Cyp17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007809.3 | NP_031835.3 | 5 - 6 | 1122 | 79 | |
| GenBank mRNA | AK132830.1 | - | 5 - 6 | 1016 | 79 | |
| AK134376.1 | - | 5 - 6 | 1016 | 79 | ||
| M64863.1 | - | 5 - 6 | 995 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp17a1
Genomic Map