Assay Details
| Gene Symbol: Mpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007863.2 | NP_031889.2 | 8 - 9 | 802 | 65 | |
| GenBank mRNA | AF079366.1 | - | 8 - 9 | 753 | 65 | |
| AK032270.1 | - | 7 - 8 | 2076 | 65 | ||
| AY236514.1 | - | 7 - 8 | 636 | 65 | ||
| BC052003.1 | - | 1049 | 65 | |||
| BC062642.1 | - | 8 - 9 | 770 | 5718318 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpp3
Genomic Map