Assay Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007995.3 | NP_032021.1 | 1 - 2 | 225 | 116 | |
| GenBank mRNA | AB007813.1 | - | 1 - 2 | 117 | 116 | |
| AK133539.1 | - | 1 - 2 | 202 | 116 | ||
| AK138752.1 | - | 1 - 2 | 172 | 116 | ||
| AK159651.1 | - | 1 - 2 | 195 | 116 | ||
| BC019180.1 | - | 1 - 2 | 173 | 5008934 | 116 | |
| CT010383.1 | - | 1 - 2 | 87 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcna
Genomic Map