Assay Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007995.3 | NP_032021.1 | 2 - 3 | 339 | 71 | |
| GenBank mRNA | AB007813.1 | - | 2 - 3 | 231 | 71 | |
| AK133539.1 | - | 2 - 3 | 316 | 71 | ||
| AK138752.1 | - | 2 - 3 | 286 | 71 | ||
| AK159651.1 | - | 2 - 3 | 309 | 71 | ||
| BC019180.1 | - | 2 - 3 | 287 | 5008934 | 71 | |
| CT010383.1 | - | 2 - 3 | 201 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcna
Genomic Map