Assay Details
| Gene Symbol: Fcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007995.3 | NP_032021.1 | 5 - 6 | 466 | 112 | |
| GenBank mRNA | AB007813.1 | - | 5 - 6 | 358 | 112 | |
| AK133539.1 | - | 5 - 6 | 443 | 112 | ||
| AK138752.1 | - | 5 - 6 | 413 | 112 | ||
| AK159651.1 | - | 5 - 6 | 436 | 112 | ||
| BC019180.1 | - | 5 - 6 | 414 | 5008934 | 112 | |
| CT010383.1 | - | 5 - 6 | 328 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcna
Genomic Map