| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900014414.mm.1 |
NM_001164717.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014414.mm.1 |
NM_001164717.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014414.mm.1 |
NM_008018.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014414.mm.1 |
NM_008018.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014414.mm.1 |
TC1900001630.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014417.mm.1 |
NM_001164717.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014417.mm.1 |
NM_001164717.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014417.mm.1 |
NM_008018.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014417.mm.1 |
NM_008018.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014417.mm.1 |
TC1900001630.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014413.mm.1 |
NM_001164717.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014413.mm.1 |
NM_001164717.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014413.mm.1 |
NM_008018.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014413.mm.1 |
NM_008018.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014413.mm.1 |
TC1900001630.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014416.mm.1 |
NM_001164717.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014416.mm.1 |
NM_001164717.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014416.mm.1 |
NM_008018.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014416.mm.1 |
NM_008018.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014416.mm.1 |
TC1900001630.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014412.mm.1 |
NM_001164717.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014412.mm.1 |
NM_001164717.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014412.mm.1 |
NM_008018.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014412.mm.1 |
NM_008018.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014412.mm.1 |
TC1900001630.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900014415.mm.1 |
NM_001164717.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014415.mm.1 |
NM_001164717.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014415.mm.1 |
NM_008018.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014415.mm.1 |
NM_008018.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900014415.mm.1 |
TC1900001630.mm.1 |
MTA TC |