Assay Details
| Gene Symbol: Sh3pxd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008018.4 | NP_032044.2 | 10 - 11 | 1076 | 59 | |
| GenBank mRNA | AJ007012.1 | - | 10 - 11 | 1076 | 59 | |
| AK085227.1 | - | 3 - 4 | 915 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3pxd2a
Genomic Map