Assay Details
| Gene Symbol: Sh3pxd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164717.1 | NP_001158189.1 | 13 - 14 | 1617 | 75 | |
| NM_008018.4 | NP_032044.2 | 14 - 15 | 1701 | 75 | ||
| GenBank mRNA | AJ007012.1 | - | 14 - 15 | 1701 | 75 | |
| AK085227.1 | - | 7 - 8 | 1540 | 75 | ||
| BC118022.1 | - | 13 - 14 | 1453 | 40097448 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3pxd2a
Genomic Map