Assay Details
| Gene Symbol: Smad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008539.3 | NP_032565.2 | 3 - 4 | 1056 | 60 | |
| GenBank mRNA | AK017583.1 | - | 3 - 4 | 1017 | 60 | |
| AK054104.1 | - | 3 - 4 | 1017 | 60 | ||
| AK084505.1 | - | 3 - 4 | 1124 | 60 | ||
| BC058693.1 | - | 3 - 4 | 1056 | 6811514 | 60 | |
| U58992.1 | - | 2 - 3 | 729 | 60 | ||
| U74359.1 | - | 3 - 4 | 922 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smad1
Genomic Map