Assay Details
| Gene Symbol: Man2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008549.2 | NP_032575.2 | 8 - 9 | 1460 | 95 | |
| GenBank mRNA | AK144763.1 | - | 2375 | 95 | ||
| BC138371.1 | - | 8 - 9 | 1445 | 8861391 | 95 | |
| BC138372.1 | - | 8 - 9 | 1445 | 8861392 | 95 | |
| X61172.1 | - | 8 - 9 | 1460 | 95 | ||
More Information
Set Membership: |
|

Man2a1
Genomic Map