Assay Details
| Gene Symbol: Sypl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008596.1 | NP_032622.1 | 4 - 5 | 636 | 82 | |
| GenBank mRNA | AB010140.1 | - | 4 - 5 | 636 | 82 | |
| AK082471.1 | - | 4 - 5 | 655 | 82 | ||
| AK083422.1 | - | 4 - 5 | 686 | 82 | ||
| AK162663.1 | - | 4 - 5 | 666 | 82 | ||
| BC030038.1 | - | 4 - 5 | 619 | 1068035 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sypl2
Genomic Map