Assay Details
| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008625.2 | NP_032651.2 | 6 - 7 | 1167 | 113 | |
| GenBank mRNA | BC141338.1 | - | 6 - 7 | 1119 | 9055950 | 113 |
| Z11974.1 | - | 6 - 7 | 1167 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrc1
Genomic Map