Assay Details
| Gene Symbol: Mthfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008638.2 | NP_032664.1 | 3 - 4 | 487 | 66 | |
| GenBank mRNA | AK076019.1 | - | 3 - 4 | 440 | 66 | |
| AK076801.1 | - | 3 - 4 | 437 | 66 | ||
| AK152249.1 | - | 3 - 4 | 479 | 66 | ||
| AK159680.1 | - | 3 - 4 | 460 | 66 | ||
| AK165840.1 | - | 3 - 4 | 479 | 66 | ||
| AK166262.1 | - | 3 - 4 | 470 | 66 | ||
| BC019511.1 | - | 3 - 4 | 474 | 4164253 | 66 | |
| J04627.1 | - | 3 - 4 | 430 | 66 | ||
| Gene Symbol: Gsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK199322.1 | - | 38 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mthfd2
Genomic Map