Assay Details
| Gene Symbol: Mthfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008638.2 | NP_032664.1 | 6 - 7 | 844 | 75 | |
| GenBank mRNA | AK076019.1 | - | 6 - 7 | 797 | 75 | |
| AK152249.1 | - | 6 - 7 | 836 | 75 | ||
| AK159680.1 | - | 6 - 7 | 817 | 75 | ||
| AK165840.1 | - | 6 - 7 | 836 | 75 | ||
| AK166262.1 | - | 6 - 7 | 828 | 75 | ||
| BC019511.1 | - | 6 - 7 | 831 | 4164253 | 75 | |
| J04627.1 | - | 6 - 7 | 787 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mthfd2
Genomic Map