Assay Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008767.2 | NP_032793.2 | 1 - 2 | 88 | 63 | |
| GenBank mRNA | AF028739.1 | - | 1 - 2 | 46 | 63 | |
| AF037065.1 | - | 1 - 2 | 59 | 63 | ||
| AJ404609.1 | - | 1 - 2 | 122 | 63 | ||
| BB869719.1 | - | 1 - 2 | 88 | 63 | ||
| BC015078.1 | - | 1 - 2 | 73 | 4224844 | 63 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF028739.1,AF037065.1,BC015078.1,NM_008767.2 transcripts.
Set Membership: |
|

Slc22a18
Genomic Map