Assay Details
| Gene Symbol: Rad51b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252562.1 | NP_001239491.1 | 3 - 4 | 253 | 92 | |
| NM_009014.3 | NP_033040.2 | 3 - 4 | 271 | 92 | ||
| GenBank mRNA | AK015771.1 | - | 3 - 4 | 253 | 92 | |
| AK032913.1 | - | 3 - 4 | 272 | 92 | ||
| U92068.1 | - | 2 - 3 | 213 | 92 | ||
More Information
Set Membership: |
|

Rad51b
Genomic Map