Assay Details
| Gene Symbol: Rapsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009023.3 | NP_033049.2 | 1 - 2 | 367 | 62 | |
| GenBank mRNA | AK009672.1 | - | 1 - 2 | 368 | 62 | |
| BC111863.1 | - | 1 - 2 | 323 | 40046186 | 62 | |
| BY709325.1 | - | 1 - 2 | 368 | 62 | ||
| J03962.1 | - | 1 - 2 | 303 | 62 | ||
| X15788.1 | - | 1 - 2 | 356 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rapsn
Genomic Map