Assay Details
| Gene Symbol: Rapsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009023.3 | NP_033049.2 | 3 - 4 | 861 | 55 | |
| GenBank mRNA | BC111863.1 | - | 3 - 4 | 817 | 40046186 | 55 |
| J03962.1 | - | 3 - 4 | 797 | 55 | ||
| X15788.1 | - | 3 - 4 | 850 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rapsn
Genomic Map