Assay Details
| Gene Symbol: Rapsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009023.3 | NP_033049.2 | 5 - 6 | 1084 | 57 | |
| GenBank mRNA | AK009672.1 | - | 4 - 5 | 928 | 57 | |
| BC111863.1 | - | 5 - 6 | 1040 | 40046186 | 57 | |
| J03962.1 | - | 5 - 6 | 1020 | 57 | ||
| X15788.1 | - | 5 - 6 | 1073 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rapsn
Genomic Map