Assay Details
| Gene Symbol: Rapsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009023.3 | NP_033049.2 | 6 - 7 | 1136 | 61 | |
| GenBank mRNA | AK009672.1 | - | 5 - 6 | 980 | 61 | |
| AK213318.1 | - | 2 - 3 | 72 | 61 | ||
| BC111863.1 | - | 6 - 7 | 1092 | 40046186 | 61 | |
| J03962.1 | - | 6 - 7 | 1072 | 61 | ||
| X15788.1 | - | 6 - 7 | 1125 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rapsn
Genomic Map