Assay Details
| Gene Symbol: Rbpjl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009036.1 | NP_033062.1 | 6 - 7 | 716 | 78 | |
| GenBank mRNA | AK131814.1 | - | 6 - 7 | 697 | 78 | |
| BC152311.1 | - | 6 - 7 | 710 | 40090714 | 78 | |
| Y10926.1 | - | 6 - 7 | 716 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbpjl
Genomic Map