Assay Details
| Gene Symbol: Rcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009037.2 | NP_033063.1 | 3 - 4 | 721 | 75 | |
| GenBank mRNA | AK017494.1 | - | 3 - 4 | 628 | 75 | |
| AK133971.1 | - | 3 - 4 | 671 | 75 | ||
| AK159973.1 | - | 3 - 4 | 624 | 75 | ||
| BC049108.1 | - | 3 - 4 | 614 | 6414128 | 75 | |
| D13003.1 | - | 3 - 4 | 641 | 75 | ||
More Information
Set Membership: |
|

Rcn1
Genomic Map