Assay Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009105.4 | NP_033131.2 | 6 - 7 | 707 | 69 | |
| GenBank mRNA | AK009288.1 | - | 6 - 7 | 600 | 69 | |
| AK011857.1 | - | 6 - 7 | 600 | 69 | ||
| AK013076.1 | - | 2 - 3 | 178 | 69 | ||
| AK014749.1 | - | 1 - 2 | 50 | 69 | ||
| AK135491.1 | - | 7 - 8 | 690 | 69 | ||
| AK149835.1 | - | 6 - 7 | 577 | 69 | ||
| AK150011.1 | - | 6 - 7 | 575 | 69 | ||
| AK152655.1 | - | 6 - 7 | 587 | 69 | ||
| AK169076.1 | - | 6 - 7 | 625 | 69 | ||
| BC003827.1 | - | 6 - 7 | 570 | 3493122 | 69 | |
| CX228720.1 | - | 6 - 7 | 707 | 69 | ||
| X63039.1 | - | 6 - 7 | 522 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rsu1
Genomic Map