Assay Details
| Gene Symbol: Atxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009125.2 | NP_033151.2 | 5 - 6 | 985 | 84 | |
| GenBank mRNA | AF041472.1 | - | 5 - 6 | 984 | 84 | |
| U70670.1 | - | 5 - 6 | 527 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn2
Genomic Map