Assay Details
| Gene Symbol: Atxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009125.2 | NP_033151.2 | 8 - 9 | 1398 | 118 | |
| GenBank mRNA | AF041472.1 | - | 8 - 9 | 1397 | 118 | |
| AK135976.1 | - | 3 - 4 | 355 | 118 | ||
| U70670.1 | - | 8 - 9 | 940 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atxn2
Genomic Map