Assay Details
| Gene Symbol: Frrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113478.1 | NP_001106950.1 | 9 - 10 | 1188 | 105 | |
| NM_009146.3 | NP_033172.2 | 9 - 10 | 1150 | 105 | ||
| GenBank mRNA | AK075573.1 | - | 9 - 10 | 1142 | 105 | |
| AK144933.1 | - | 9 - 10 | 1142 | 105 | ||
| AK162693.1 | - | 9 - 10 | 1143 | 105 | ||
| AK167289.1 | - | 9 - 10 | 1178 | 105 | ||
| BC027770.1 | - | 8 - 9 | 1148 | 5344922 | 105 | |
| D50464.1 | - | 9 - 10 | 1138 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frrs1
Genomic Map